
Manganese(II) bromide tetrahydrate, Sigma-Aldrich®
Catalog # SIAL208434-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:286.81 g/mol
:M-Clarity™ quality level = MQ100
EINECS:236-591-9
:GHS07
Storage Temperature:Ambient
CAS Number:10031-20-6
Formula:Br₂Mn·4H₂O
MDL Number:MFCD00149791