


Chromium(III) nitrate nonahydrate, EMSURE® for analysis, Supelco®
Catalog # 1.02481.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:810 g/l (20 °C)
- Melting Point:36 - 37 °C
- Molar Mass:400.15 g/mol
- Bulk Density:710 kg/m3
- pH:2 - 3 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:400.15 g/mol
Merck Index:13,02246
Melting Pt:60 °C
Density:1.807 g/cm³ (20 °C)
EINECS:236-921-1
UN:2720
ADR:5.1,III
:M-Clarity™ quality level = MQ300
:GHS03
:GHS07
:GHS09
CAS Number:7789-02-8
Formula:CrN₃O₉·9H₂O
MDL Number:MFCD00149659