
Yttrium(III) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-94-2
:
Formula:YCl₃·6H₂O
MDL Number:MFCD00149941
:303.36 g/mol
Boiling Pt:1507 °C (1013 hPa)
Melting Pt:153 °C
Density:2.18 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:233-801-0
:GHS07
Frequently Bought Together