


Mercury(II) nitrate monohydrate ACS
:7783-34-8
:13465-31-1
:Hg(NO₃)₂·1H₂O
:MFCD00149736
:342.62 g/mol
:77…79 °C
:4.3 g/cm³
:233-152-3
:1625
:6.1,II
:GHS06
:GHS08
:GHS09
:Ambient
- Appearance:White to off-white Crystalline powder or crystals
- Titration with NH4SCN:≥98.0 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤20 ppm
- Iron (Fe):≤10 ppm
- Residue after reduction:≤0.01 %
Frequently Bought Together