


Mercury(II) nitrate monohydrate 98+%
:7783-34-8
:13465-31-1
:Hg(NO₃)₂·1H₂O
:MFCD00149736
:342.62 g/mol
:77…79 °C
:4.3 g/cm³
:233-152-3
:1625
:6.1,II
:GHS06
:GHS08
:GHS09
:Ambient
- Appearance:White Adhering crystalline powder
- Titration Complexometric:≥98 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤20 ppm
- Copper (Cu):≤20 ppm
- Iron (Fe):≤20 ppm
- Lead (Pb):≤20 ppm
- Mercurous Mercury (as Hg):≤0.2 %
Frequently Bought Together