Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
 
 
 
Dusičnan rtuťný dihydrát 98% p.a.
 
undefined
 
 
 
Dusičnan rtuťný dihydrát 98% p.a.
 

 

  • Appearance:
    White to yellow Crystalline powder
  • Assay:
    ≥97.5 %
  • Chloride (Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.005 %
  • Iron (Fe):
    ≤0.001 %
  • Mercuric Mercury (as Hg):
    ≤0.5 %
  • Insoluble matter:
    ≤0.005 % (in dilute HNO3)
  • Residue after reduction:
    ≤0.01 %

 

 

Frequently Bought Together