


Cadmium(II) nitrate tetrahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10022-68-1
:
Formula:CdN₂O₆·4H₂O
MDL Number:MFCD00149626
:308.48 g/mol
Merck Index:14,1622
Boiling Pt:132 °C
Melting Pt:59 °C
Density:2.45
Storage Temperature:Ambient
EINECS:233-710-6
UN:3087
ADR:5.1,II
:GHS06
:GHS08
:GHS03
Frequently Bought Together