

Cadmium(II) bromide tetrahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13464-92-1
:
Formula:Br₂Cd·4H₂O
MDL Number:MFCD00150204
:344.28 g/mol
Melting Pt:575 °C
Density:5.214 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-165-1
UN:2570
ADR:6.1,III
:GHS07
:GHS09
Related Information
Frequently Bought Together