IHC fluoromount, HIGHDEF®
Catalog # ENZOADI9502600025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 mL
- Description:IHC fluoromount
- Seq:02
Specifications
About this item
Fluoromount is an aqueous-based permanent, high stability mounting medium designed for fluorogenic labelling and ideal for long-term storage of processed tissue.
- High definition visualisation for IHC, IF, ISH, FISH
- It is compatible with FITC, phycoerythrin, phycocyanin, and allophycocyanin
- Can also be used with other applications
Formulation: Liquid, mounting medium containing <0,1% sodium azide.