Order Entry
Czech Republic
ContactUsLinkComponent
Copper(II) chloride dihydrate ≥96%, TECHNICAL
Irritant
Environment
Copper(II) chloride dihydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Environment
Copper(II) chloride dihydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results

Frequently Bought Together