Copper(II) chloride dihydrate 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate
CAS Number:10125-13-0
:
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674
Density:2.510
Molecular Weight:170.48 g/mol
Melting Pt:100 °C (dec.)
ADR:8 III
Merck Index:14,2633
EINECS:231-210-2
UN:2802
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together