

Copper(II) chloride dihydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10125-13-0
:
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674
:170.48 g/mol
Merck Index:14,2633
Melting Pt:100 °C (dec.)
Density:2.510
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together