

Samarium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13759-83-6
:24581-35-9
Formula:N₃O₉Sm·6H₂O
MDL Number:MFCD00149857
:444.47 g/mol
Melting Pt:78…79 °C
Density:2.375
Storage Temperature:Ambient
EINECS:233-798-6
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together