

Lanthanum(III) nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10277-43-7
:10227-43-7
Formula:La(NO₃)₃·6H₂O
MDL Number:MFCD00149751
:433.01 g/mol
Merck Index:14,05363
Boiling Pt:126 °C (1013 hPa)
Melting Pt:65…67 °C
Density:2.347 g/cm³ (20 °C)
UN:1477
ADR:5.1,III
:M-Clarity™ quality level = MQ200
EINECS:233-238-0
:GHS03
:GHS07
Storage Temperature:Ambient
Frequently Bought Together