

Holmium trinitrate pentahydrate, Sigma-Aldrich®
Catalog # SIAL229687-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:441.02 g/mol
Melting Pt:83.74…84.62 °C
UN:1477
ADR:5.1,III
:M-Clarity™ quality level = MQ100
EINECS:233-438-8
:GHS03
:GHS07
CAS Number:14483-18-2
Formula:HoN₃O₉·5H₂O
MDL Number:MFCD00149737