

Holmium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14483-18-2
:
Formula:HoN₃O₉·5H₂O
MDL Number:MFCD00149737
:441.02 g/mol
Melting Pt:83.74…84.62 °C
Storage Temperature:Ambient
EINECS:233-438-8
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together