Order Entry
Czech Republic
ContactUsLinkComponent
Erbium trinitrate pentahydrate 99.9% (trace metals basis)
Oxidizing
Irritant
Erbium trinitrate pentahydrate 99.9% (trace metals basis)
Certificates
undefined
Oxidizing
Irritant
Erbium trinitrate pentahydrate 99.9% (trace metals basis)

Specification Test Results

  • Appearance:
    Pink Crystalline powder or crystals
  • Assay:
    33 to 39 % (Er)
  • Total conc. of imp.as REO:
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together