Samarium trichloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL204277-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:364.81 g/mol
Density:2.383 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
EINECS:233-797-0
CAS Number:13465-55-9
Formula:Cl₃Sm·6H₂O
MDL Number:MFCD00011235