
Holmium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14914-84-2
:
Formula:Cl₃Ho·6H₂O
MDL Number:MFCD00149739
:379.38 g/mol
Merck Index:14,4727
Melting Pt:718 °C
EINECS:233-387-1
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together