
Gadolinium trichloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13450-84-5
:
Formula:Cl₃Gd·6H₂O
MDL Number:MFCD00149732
:371.7 g/mol
:M-Clarity™ quality level = MQ100
EINECS:233-386-6
:GHS07
Frequently Bought Together