Europium(III) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13759-92-7
:
Formula:Cl₃Eu·6H₂O
MDL Number:MFCD00149701
:366.41 g/mol
Melting Pt:158 °C
Density:4.89 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:233-040-4
Frequently Bought Together