
Ammonium iron(III) sulphate dodecahydrate 98.5-102% ACS
Catalog # 0232-500G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Calcium:<0.01%
- Chloride:≤0.001%
- Copper:≤0.003%
- Ferrous Iron:≤0.001%
- Insolubles:≤0.01%
- Magnesium:≤0.005%
- Nitrate:≤0.01%
- Potassium:≤0.005%
- Purity:98.5 – 102%
- Sodium:≤0.02%
- Zinc:≤0.003%
Specification Test Results
About this item
:
:482.2 g/mol
Merck Index:12,00549
Boiling Pt:230 °C (1013 hPa)
Melting Pt:39…41 °C
Density:1.71 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:233-382-4
:GHS07
CAS Number:7783-83-7
Formula:NH₄Fe(SO₄)₂·12H₂O
MDL Number:MFCD00150004