

Tin(II) chloride dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:231-868-0
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together