
Chlorid hlinitý hexahydrát, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7784-13-6
:
Formula:AlCl₃·6H₂O
MDL Number:MFCD00149134
Melting Pt:∼100 °C
Density:2,39 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-208-1
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS07
Frequently Bought Together