Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Aluminium chloride hexahydrate 99%, Reagent Grade
 
 
Aluminium chloride hexahydrate 99%, Reagent Grade
 

 

  • Vapor Pressure:
    1 mmHg ( 100 °C)
  • Assay:
    98.5 - 101.5 %

 

 

Frequently Bought Together