Beryllium sulphate tetrahydrate ≥99.99% (metals basis)
:7787-56-6
:
:BeO₄S·4H₂O
:MFCD00149156
:580
:110
:1.1713
:177.14 g/mol
:Ambient
:236-842-2
:6.1 II
:14,1175
:1566
:GHS06
:GHS08
:GHS07
- Environmentally Preferable:
Frequently Bought Together