

Barium hydroxide octahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.01737.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Solubility:72 g/l (20 °C)
- Melting Point:78 °C
- Molar Mass:315.48 g/mol
- Bulk Density:900 - 1100 kg/m3
- Density:2.18 g/cm3 (20 °C)
- pH:12.5 (50 g/l, H2O, 20 °C) (slurry)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:315.47 g/mol
Merck Index:13,00980
Boiling Pt:780 °C (1013 hPa)
Melting Pt:78 °C
Density:2.18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:241-234-5
UN:2923
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ300
:GHS05
:GHS07
CAS Number:12230-71-6
Formula:Ba(OH)₂·8H₂O
MDL Number:MFCD00149152