
Strontium chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-70-4
:
Formula:SrCl₂·6H₂O
MDL Number:MFCD00149865
:266.62 g/mol
Merck Index:13,08920
Boiling Pt:100 °C (1013 hPa)
Melting Pt:61 °C
Density:1.93 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:233-971-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together