Order Entry
Czech Republic
ContactUsLinkComponent
Magnesium chloride hexahydrate 98.0-101.0% Ph. Eur., USP, BP
Magnesium chloride hexahydrate 98.0-101.0% Ph. Eur., USP, BP
Supplier:  VWR Chemicals
Certificates
undefined
Magnesium chloride hexahydrate 98.0-101.0% Ph. Eur., USP, BP
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Colourless crystals
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • Residual solvents:
    Unlikely by manuf.process
  • Residues of metal catalysts or reagents:
    Unlikely by manuf.process
  • As (Arsenic):
    ≤ 2 ppm
  • Cd (Cadmium):
    ≤ 1 ppm
  • Co (Cobalt):
    ≤ 1 ppm
  • Hg (Mercury):
    ≤ 2 ppm
  • Ni (Nickel):
    ≤ 6 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • V (Vanadium):
    ≤ 10 ppm
  • Conforms to Ph.Eur./USP/BP:
    Passes test
  • Ph.Eur./BP specifications::
    .
  • Assay [Ph.Eur./BP]:
    98.0 - 101.0 %
  • Identification A:
    Passes test Ph.Eur./BP
  • Identification B:
    Passes test Ph.Eur./BP
  • Identification C:
    Passes test Ph.Eur./BP
  • Appearance of solution:
    Passes test Ph.Eur./BP
  • Acidity or alkalinity:
    Passes test Ph.Eur./BP
  • Br (Bromide) [Ph.Eur./BP]:
    ≤ 500 ppm
  • SO4 (Sulphate) [Ph.Eur./BP]:
    ≤ 100 ppm
  • Ca (Calcium) [Ph.Eur./BP]:
    ≤ 0.1 %
  • Fe (Iron) [Ph.Eur./BP]:
    ≤ 10 ppm
  • Water [Ph.Eur./BP]:
    51.0 - 55.0 %
  • USP specifications::
    .
  • Assay [USP]:
    98.0 - 101.0 %
  • Insoluble matter [USP]:
    ≤ 0.005 %
  • SO4 (Sulphate) [USP]:
    ≤ 0.005 %
  • Ba (Barium):
    Passes test USP
  • Limit of calcium [USP]:
    ≤ 0.01 %
  • K (Potassium):
    Passes test USP
  • pH (25°C; 50 mg/ml) [USP]:
    4.5 - 7.0

Specification Test Results

Frequently Bought Together