
Barium chloride dihydrate, Sigma-Aldrich®
Catalog # SIAL529591-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:244.27 g/mol
Merck Index:14,00971
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:960 °C
Density:3.86 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:1564
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS06
CAS Number:10326-27-9
Formula:BaCl₂·2H₂O
MDL Number:MFCD00149154