Order Entry
Czech Republic
ContactUsLinkComponent
Barium chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
Toxic
Barium chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Toxic
Barium chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Calcium (Ca):
    max. 0.05 %
  • Iron (Fe):
    max. 2 ppm
  • Potassium (K):
    max. 0.0025 %
  • Sodium (Na):
    max. 0.005 %
  • Strontium (Sr):
    max. 0.1 %
  • Heavy Metals (as Pb):
    max. 5 ppm
  • Insoluble Matter:
    max. 0.005 %
  • Loss on Drying at 150°C:
    14.0 - 16.0 %
  • Oxidizing Substances (as NO3):
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    5.2 - 8.2

Specification Test Results

Related Information

Frequently Bought Together