Order Entry
Czech Republic
ContactUsLinkComponent
Chromium(III) potassium sulphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Irritant
Chromium(III) potassium sulphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.01036.0250
Supplier:  Merck
CAS Number:  
Irritant
Chromium(III) potassium sulphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.01036.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    250 g/l (25 °C)
  • Melting Point:
    89 °C
  • Molar Mass:
    499.41 g/mol
  • Bulk Density:
    800 - 1000 kg/m3
  • Density:
    1.83 g/cm3 (20 °C)
  • pH:
    3 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :499.4 g/mol
Merck Index:13,02250
Melting Pt:89 °C
Density:1.83 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
EINECS:233-401-6
 :GHS07
CAS Number:7788-99-0
Formula:CrK(SO₄)₂·12H₂O
MDL Number:MFCD00149917