
Azido-PEG3-sulphorhodamine 101 conjugate
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:C₃₉H₄₆N₆O₉S₂
MDL Number:MFCD22380752
:806.95 g/mol
:GHS07
Storage Temperature:Freezer
Related Information
Frequently Bought Together