Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hexacyanoferrate(II) trihydrate, Sigma-Aldrich®
Environment
Potassium hexacyanoferrate(II) trihydrate, Sigma-Aldrich®
Catalog # SIAL60279-250G
Supplier:  Merck
CAS Number:  
Environment
Potassium hexacyanoferrate(II) trihydrate, Sigma-Aldrich®
Catalog # SIAL60279-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :422.39 g/mol
Merck Index:12,07793
Melting Pt:70 °C
Density:1.889 g/cm³ (20 °C)
EINECS:237-722-2
 :GHS09
Storage Temperature:Ambient
CAS Number:14459-95-1
Formula:K₄Fe(CN)₆·3H₂O
MDL Number:MFCD00167023