Potassium hexacyanoferrate(II) trihydrate, extra pure, Supelco®
1.04982.1000
: Merck
:
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
:
:422.39 g/mol
:12,07793
:70 °C
:1.889 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:237-722-2
:GHS09
:Ambient
:14459-95-1
:K₄Fe(CN)₆·3H₂O
:MFCD00167023