Order Entry
Czech Republic
ContactUsLinkComponent
tetra-Chloroauric(III) acid trihydrate ACS
Corrosive
Irritant
tetra-Chloroauric(III) acid trihydrate ACS
Catalog # 0240-5G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
tetra-Chloroauric(III) acid trihydrate ACS
Catalog # 0240-5G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Alkalies and Other Metals:
    <0.2%
  • Insoluble in Ether:
    ≤0.1%
  • Purity (Au):
    ≥49%

Specification Test Results

About this item

 :
 :393.83 g/mol
Melting Pt:30 °C
Density:3.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-948-4
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:16961-25-4
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904