Order Entry
Czech Republic
ContactUsLinkComponent
p-Toluenesulphonic acid monohydrate, EMSURE® ACS for analysis, Supelco®
Irritant
p-Toluenesulphonic acid monohydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates
Irritant
p-Toluenesulphonic acid monohydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    670 g/l (20 °C)
  • Melting Point:
    56 °C Elimination of water of crystallisation
  • Molar Mass:
    190.22 g/mol
  • Bulk Density:
    510 kg/m3
  • Boiling Point:
    140 °C (27 hPa)
  • Flash Point:
    180 °C
  • pH:
    1 (650 g/l, H2O, 20 °C) (anhydrous substance)
  • Ignition Point:
    600 °C

Specification Test Results

Related Information

Frequently Bought Together