EDTA disodium salt dihydrate 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
Specification Test Results
- Assay:99.0 - 101.0 %
- pH (25°C; 5 %):4.0 - 5.5
- Identification (IR):Passes test
- Identification B:Passes test Ph.Eur.
- Identification (Na):Passes test
- Appearance of Solution S:Passes test Ph.Eur.
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Loss on drying (150°C):9 - 10 %
- Nitrilotriacetic acid:Max. 0.05 %
- Cl (Chloride):Max. 40 ppm
- CN (Cyanide):Max. 10 ppm
- SO4 (Sulphate):Max. 0.02 %
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 5 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 50L Plastic bucket UN Y (ZPACK13247050) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
20302_EDTA DISODIUM SALT 2H2O ANALAR R.PEACS_BSE_REV0.pdf
BSE/TSE Certificates
SDS_EN_CH_VWRC20302.pdf SDS_EN_DE_VWRC20302.pdf SDS_EN_NL_VWRC20302.pdf SDS_EN_UK_VWRC20302.pdf sds_VWRC20302
SDS
spec_VWRC20302.180 spec_VWRC20302.180 spec_VWRC20302.236 spec_VWRC20302.236 spec_VWRC20302.260
Product specification
More
Frequently Bought Together