Order Entry
Czech Republic
ContactUsLinkComponent
EDTA disodium salt dihydrate ≥98%, TECHNICAL
EDTA disodium salt dihydrate ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
EDTA disodium salt dihydrate ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

Frequently Bought Together