Order Entry
Czech Republic
ContactUsLinkComponent
EDTA disodium salt dihydrate 99+% ACS
Irritant
Health
EDTA disodium salt dihydrate 99+% ACS
Certificates
undefined
Irritant
Health
EDTA disodium salt dihydrate 99+% ACS
Certificates

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Complexometric:
    99.0 to 101.0 %
  • pH:
    4.0 to 6.0 (5 % at 25°C)
  • Iron (Fe):
    ≤100 ppm
  • Heavy metals (as Pb):
    ≤50 ppm
  • Nitrilotriacetic acid:
    ≤0.1 %
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together