Order Entry
Czech Republic
ContactUsLinkComponent
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Irritant
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Catalog # 0116-07
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Catalog # 0116-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • GMP Manufactured Product:
    99.5 - 100.5%
  • Meets B.P. Chemical Specifications:
    Passes Test
  • Meets E.P. Chemical Specifications:
    Passes Test
  • Meets J.P. Chemical Specifications:
    Passes Test
  • Meets U.S.P Requirements:
    ≤0.036%
  • Preserve in Tight Containers:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.1%
  • Bulk Pharmaceutical Chemical:
    ≤0.015%
  • USP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • USP - Clarity of Solution:
    Passes Test
  • USP - Color of Solution:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Oxalic Acid:
    ≤ 0.036 %
  • USP - Readily Carbonizable Substances:
    Passes Test
  • USP - Residue on Ignition:
    ≤ 0.1 %
  • USP - Sulfate (SO₄):
    ≤ 0.015 %
  • USP - Water (H₂O):
    7.5 - 9.0 %
  • EP/BP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • EP/BP - Endotoxin Concentration, IU/mg:
    ≤ 0.5
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification E:
    Passes Test
  • EP/BP - Oxalic Acid:
    ≤ 360 ppm
  • EP/BP - Readily Carbonizable Substances:
    Passes Test
  • EP/BP - Sulfate (SO₄):
    ≤ 150 ppm
  • EP/BP - Water (H₂O):
    7.5 - 9.0 %
  • JP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):
    99.5 - 100.5 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Identification:
    Passes Test
  • JP - Oxalic Acid:
    ≤ 360 ppm
  • JP - Water (H₂O):
    7.5 - 9.0 %
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Residue on Ignition:
    ≤ 0.10 %
  • JP - Sulfate (SO₄):
    ≤ 150 ppm
  • ACS - Iron (Fe):
    ≤ 3 ppm
  • Lead (Pb):
    ≤ 2 ppm

Specification Test Results

About this item

 :
 :210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.54 g/cm³
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
 :GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972