di-Ammonium oxalate monohydrate ≥99.5% (RT), BioUltra, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6009-70-7
:
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694
:142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:∼1.5 g/cm³ (20 °C)
UN:3288
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:214-202-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together