Order Entry
Czech Republic
ContactUsLinkComponent
Oxalic acid dihydrate, Suprapur® for trace analysis, Supelco®
Oxalic acid dihydrate, Suprapur® for trace analysis, Supelco®
Catalog # 1.00489.0100
Supplier:  Merck
CAS Number:  
undefined
Oxalic acid dihydrate, Suprapur® for trace analysis, Supelco®
Catalog # 1.00489.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    102 g/l (20 °C)
  • Melting Point:
    101 °C
  • Molar Mass:
    126.07 g/mol
  • Bulk Density:
    900 kg/m3
  • Boiling Point:
    149 - 160 °C (decomposition)
  • Vapor Pressure:
    21 hPa (50 °C)
  • Flash Point:
    157 °C (decomposition)
  • Density:
    1.65 g/cm3 (20 °C)
  • pH:
    1 (10 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

Synonyms: Ethanedioic acid dihydrate
 :
 :126.07 g/mol
EINECS:205-634-3
UN:3261
REACH:01-2119534576-33
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:6153-56-6
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102
Boiling Pt:149…160 °C (1013 hPa)
Melting Pt:101
Density:1.65 g/cm³ (20 °C)
Merck Index:13,06980
 :M-Clarity™ quality level = MQ100