


Nickel acetate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6018-89-9
:
Formula:C₄H₆NiO₄·4H₂O
MDL Number:MFCD00066973
:248.84 g/mol
Merck Index:12,06583
Boiling Pt:16 °C (1013 hPa)
Density:1.798 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:206-761-7
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together