


Nickel acetate tetrahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6018-89-9
:
Formula:C₄H₆NiO₄·4H₂O
MDL Number:MFCD00066973
:248.84 g/mol
Merck Index:14,6499
Boiling Pt:16 °C (1013 hPa)
Melting Pt: Dec.
Density:1.744
Storage Temperature:Ambient
EINECS:206-761-7
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Related Information
Frequently Bought Together