Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Nickel acetate tetrahydrate 99%, extra pure
 
undefined
 
 
 
Nickel acetate tetrahydrate 99%, extra pure

 

  • Appearance:
    Green to blue Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥98.5 %
  • Cobalt (Co):
    ≤0.2 %
  • Iron (Fe):
    ≤20 ppm

 

 

Frequently Bought Together