

Cadmium acetate dihydrate ≥98.0% (by complexometric analysis), Purum p.a.
Catalog # 20901-500G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:98.0 - 102.0 %
- Calcium (Ca):Max. 50 ppm
- Copper (Cu):Max. 50 ppm
- Iron (Fe):Max. 50 ppm
- Sodium (Na):Max. 100 ppm
- Lead (Pb):Max. 50 ppm
- Zinc (Zn):Max. 50 ppm
- Total S (as SO₄):Max. 50 ppm
- Chloride (Cl):Max. 50 ppm
- Sulfate (SO₄):Max. 50 ppm
Specification Test Results
About this item
:
:266.53 g/mol
Merck Index:13,01614
Boiling Pt: Dec.
Melting Pt:256 °C.anhydrous substance.Elimination of water of crystallisation
Melting Pt:256 °C (anhy) (–H₂O of crystall.)
Density:2.01 g/cm³ (20 °C)
EINECS:208-853-2
ADR:6.1,III
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:5743-04-4
Formula:(H₃CCOO)₂Cd·2H₂O
MDL Number:MFCD00150020