

Cadmium acetate dihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5743-04-4
:
Formula:(H₃CCOO)₂Cd·2H₂O
MDL Number:MFCD00150020
:266.53 g/mol
Merck Index:14,1614
Boiling Pt:105…107 °C (4 mmHg)
Melting Pt:256 °C
Density:2.01
Storage Temperature:Ambient
EINECS:208-853-2
UN:2570
ADR:6.1,III
:GHS07
:GHS09
Related Information
Frequently Bought Together