

Lead(II) acetate trihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6080-56-4
:
Formula:(H₃CCOO)₂Pb·3H₂O
MDL Number:MFCD00150023
:379.34 g/mol
Merck Index:13,05415
Boiling Pt:280 °C (1013 hPa)
Melting Pt:75 °C
Density:2.55 g/cm³ (20 °C)
UN:1616
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:206-104-4
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together