Order Entry
Czech Republic
ContactUsLinkComponent
Magnesium acetate tetrahydrate ≥98% ACS, Fluka™
Magnesium acetate tetrahydrate ≥98% ACS, Fluka™
Certificates
Magnesium acetate tetrahydrate ≥98% ACS, Fluka™

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Insoluble in acetic acid:
    Max. 0.005 %
  • Barium (Ba):
    Max. 0.001 %
  • Calcium (Ca):
    Max. 0.01 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.005 %
  • Manganese (Mn):
    Max. 0.001 %
  • Sodium (Na):
    Max. 0.005 %
  • Strontium (Sr):
    Max. 0.005 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Form:
    Solid

Specification Test Results

Related Information

Frequently Bought Together